Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Manganese (III) acetate dihydrate
Manganese(III) acetate dihydrate ≥97%
Manganese(III) acetate dihydrate ≥97%
Irritant
Manganese(III) acetate dihydrate ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Irritant
Manganese(III) acetate dihydrate ≥97%
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
19513-05-4
:
Formula:
(H₃CCOO)₂Mn·2H₂O
MDL Number:
MFCD00150022
:
209.06 g/mol
EINECS:
213-602-5
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFA047050
Videos
Thermo Scientific Acroseal 25 Years
Documentation