N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES) ≥99.0% (by titrimetric analysis) Good's buffer component for biological research
Catalog # TCIAA0700-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White powder to crystal
- Purity (Neutralization titration):min. 99.0 %
- Absorbance:max. 0.015 (0.1mol/L、H2O、260nm)
- Solubility in hot Water:almost transparency
- Iron:max. 5 ppm
Specification Test Results
About this item
:
:182.2 g/mol
Merck Index:13,00037
Melting Pt:270…295 °C
Storage Temperature:Ambient
EINECS:230-908-4
CAS Number:7365-82-4
Formula:H₂NCOCH₂NHCH₂CH₂SO₃H
MDL Number:MFCD00008030