
5-Amino-3-methyl-1-phenyl-1H-pyrazole ≥98.0% (by HPLC, titration analysis)
Catalog # TCIAA1311-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:113.0 to 117.0 deg-C
- Solubility in Acetic acid:within almost transparency
Specification Test Results
About this item
:
:173.21 g/mol
Boiling Pt:333°C
EINECS:214-463-3
:GHS07
CAS Number:1131-18-6
Formula:C₁₀H₁₁N₃
MDL Number:MFCD00020727