(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)
Catalog # MOLE26747821-25G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:520.4 g/mol
CAS Number:128625-52-5
Formula:C₁₈H₂₈F₆N₆OP₂
MDL Number:MFCD00077411