Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bottles
Wash Bottles
Wash bottles, narrow neck, Azlon®
Wash bottles, narrow neck, Azlon®
Wash bottles, narrow neck, Azlon®
Supplier:
SciLabware
Certificates
Expand image
Wash bottles, narrow neck, Azlon®
Supplier:
SciLabware
About this item
LDPE, translucent.
Closures feature precision jet-tip aperture for fine control
Traditional and popular square shoulder round wash bottles
Related Information
Documentation
Datasheet
AZLOWGF230P_TDS_EN
Documentation