Order Entry
Finland
ContactUsLinkComponent
Hand and finger protectors, Azlon®
Hand and finger protectors, Azlon®
Catalog # AZLOPGR406
Supplier:  SciLabware
undefined
Hand and finger protectors, Azlon®
Catalog # AZLOPGR406
Supplier:  SciLabware

Specifications

  • Colour:
    White
  • Seq:
    03
  • Size:
    U*
  • Model:
    Finger protector
  • Pk:
    1

Specifications

About this item

Hand protector made of silicone rubber.

  • Ideal for safe handling of hot beakers, flasks etc
  • Capable of withstanding temperatures up to 250 °C
  • Also excellent for handling cold objects down to −57 °C
  • Contact surfaces are multi-studded for positive grip

* One size fits all.