About this item
Plasmid isolated with traditional purification procedures normally contain high levels of endotoxins (also known as lipopolysaccharides or LPS) that can significantly interfere with transfection experiments downstream. The E.Z.N.A.® Endo-Free plasmid mini kit II integrates an efficient endotoxin removal step into the plasmid purification procedure to produce high-quality transfection grade (<0,1 EU/µg) plasmid for efficient transfection. The bacterial cells are lysed using the alkaline-SDS lysis method. The cleared cell lysate is then treated with ETR reagent to efficiently remove the endotoxins. After adjusting the binding condition, the cell lysate is applied into the HiBind® DNA column and purified DNA is eluted from the column membrane.
- Endotoxins efficiently reduced (<0,1 EU/µg)
- No resin, slurries, or alcohol precipitation
- Fast, easy and convenient protocols
Specifications
- Method / Format:Column (silica-based)
- Prep size:Mini plus
- Refine target molecule:Plasmid (endotoxin-free)
- Sample type:Bacteria
- Target molecule:Plasmid
- Sample size:3 - 15 ml
- Description:E.Z.N.A.® Endo-Free Plasmid Mini Kit II