E.Z.N.A.® PX Blood RNA Kit
Isolates total RNA from blood samples stored in special preservation reagents and PAXgene tubes using spin columns.
- RNA purified using the E.Z.N.A.® PX RNA Kit method is ready for applications such as RT-PCR
Omega Bio-tek’s E.Z.N.A.® PX Blood RNA Kit is designed for isolation of total RNA from blood samples stored in special preserved reagents and Paxgene tubes. The procedure completely removes contaminants and enzyme inhibitors making RNA isolation fast, convenient, and reliable. RNA purified using the E.Z.N.A.® PX RNA Kit method is ready for applications such as RT-PCR.
- Method / Format:Column (silica-based)
- Prep size:Mini
- Refine target molecule:Total RNA
- Sample type:Blood DNA/RNA collection tubes (e.g. PAXgene )
- Target molecule:RNA