Order Entry
Finland
ContactUsLinkComponent
Malt extract
Malt extract
Catalog # COND1708
Supplier:  LABORATORIOS CONDA
CAS Number:  
Malt extract
Catalog # COND1708
Supplier:  LABORATORIOS CONDA
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
Storage Temperature:Ambient
EINECS:232-310-9
CAS Number:8002-48-0
MDL Number:MFCD00131570