Order Entry
Finland
ContactUsLinkComponent
 
 
5-Bromobenzo[b]thiophene ≥98.0% (by GC)
 :  TCI
 
undefined
 
5-Bromobenzo[b]thiophene ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    46.0 to 50.0 deg-C