Order Entry
Finland
ContactUsLinkComponent
di-Sodium tetraborate decahydrate 99.5-105.0%, granular, AR® ACS, Macron Fine Chemicals™
Irritant
Health
di-Sodium tetraborate decahydrate 99.5-105.0%, granular, AR® ACS, Macron Fine Chemicals™
Certificates
undefined
Irritant
Health
di-Sodium tetraborate decahydrate 99.5-105.0%, granular, AR® ACS, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Meets ACS Reagent Chemical Requirements:
  • For Laboratory,Research,or Manufacturing Use:
  • ACS - Assay (Na₂B₄O₇·10H₂O):
    99.5 - 105.0 %
  • ACS - pH of 0.01M Solution at 25°C:
    9.15 - 9.20
  • ACS - Insoluble Matter:
    ≤0.005 %
  • ACS - Chloride (Cl):
    ≤0.001 %
  • ACS - Phosphate (PO₄):
    ≤0.001 %
  • ACS - Sulfate (SO₄):
    ≤0.005 %
  • ACS - Calcium (Ca):
    ≤0.005 %
  • ACS - Heavy Metals (as Pb):
    ≤0.001 %
  • ACS - Iron (Fe):
    ≤5 ppm

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together