Order Entry
Finland
Orders LinkContactUsLinkComponent
Chloroform, BAKER ANALYZED® ACS, J.T.Baker®

Chloroform, BAKER ANALYZED® ACS, J.T.Baker®

Toxic
Health
Supplier:   Avantor
Chloroform, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
Chloroform, BAKER ANALYZED® ACS, J.T.Baker®

2 Options Available Below

Product Details & Documents
Synonyms:  Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
  • CAS Number:67-66-3
  • Formula:CHCl₃
  • MDL Number:MFCD00000826
  • Molecular Weight:119.38 g/mol
  • Merck Index:13,02160
  • Boiling Pt:61.7 °C (1013 hPa)
  • Melting Pt:–63 °C
  • Density:1.4832 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-663-8
  • UN:1888
  • ADR:6.1,III
  • REACH:05-2114614906-45

Documents

Specifications for Product Family

Specification Test Results

  • Assay (by GC):
    min. 99,8 %
  • Acetone and Aldehyde (as(CH3)2CO):
    max. 0,001 %
  • Acid and Chloride:
    passes test
  • Color (APHA):
    max. 10
  • Free Chlorine:
    passes test
  • Lead (Pb):
    max. 0,05 ppm
  • Preservative (C2H5OH):
    about 0.75%
  • Residue after Evaporation:
    max. 5 ppm
  • Substances Darkened by H2SO4:
    passes test
  • Suitability for Use with Dithizone Test:
    passes test
  • Ultraviolet Absorbance (1.00-cm path vs water)::
    -
  • at 245 nm:
    max. 1,00
  • at 255 nm:
    max. 0,25
  • at 260 nm:
    max. 0,15
  • at 270 nm:
    max. 0,05
  • at 290-400 nm:
    max. 0,01
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.