Order Entry
Finland
ContactUsLinkComponent
Organic reference standard, 5-tert-Butyl-2,4,6-trinitro-m-xylene 100 µg/ml in isooctane
Organic reference standard, 5-tert-Butyl-2,4,6-trinitro-m-xylene 100 µg/ml in isooctane
Catalog # PROOSL20941
CAS Number:  
Organic reference standard, 5-tert-Butyl-2,4,6-trinitro-m-xylene 100 µg/ml in isooctane
Catalog # PROOSL20941
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • CAS No.:
    81-15-2
  • Description:
    5-tert-Butyl-2,4,6-trinitro-m-xylene 100 µg/ml in isooctane
  • Formula:
    C₁₂H₁₅N₃O₆
  • Description:
    5-tert-Butyl-2,4,6-trinitro-m-xylene

Specifications

About this item