
3-Methoxy-o-toluidine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:19500-02-8
:
Formula:C₈H₁₁NO
MDL Number:MFCD06412568
ADR:6.1,III
:137.1800 g/mol
UN:2810
:GHS06
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %