Order Entry
Finland
ContactUsLinkComponent
4-Nitroaniline hydrochloride ≥99.0% (by HPLC, titration analysis)
Irritant
Health
4-Nitroaniline hydrochloride ≥99.0% (by HPLC, titration analysis)
Catalog # TCIAN0122-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
Health
4-Nitroaniline hydrochloride ≥99.0% (by HPLC, titration analysis)
Catalog # TCIAN0122-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Amber to Dark green powder to crystal
  • Purity (HPLC):
    min. 99.0 area%
  • Purity (Argentmetric Titration):
    min. 99.0 %
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :174.5850 g/mol
EINECS:239-997-4
ADR:6.1,II
 :GHS07
 :GHS08
CAS Number:15873-51-5
Formula:C₆H₇ClN₂O₂
MDL Number:MFCD00054338