
4-(N,N-Diphenylamino)phenylboronic acid (contains varying amounts of Anhydride)
: TCI
:201802-67-7
:
:C₁₈H₁₆BNO₂
:MFCD06798117
:289.14 g/mol
:228°C
:GHS07
- Appearance:White to Dark green powder to crystal
- Purity (Neutralization titration):97.0 to 108.0 %