3'-Allyl-4'-hydroxyacetophenone
Marketsource
Supplier: Thermo Scientific
Jump to:
About this item
Synonyms: 3′-Allyl-4′-hydroxyacetophenone, 1-(3-Allyl-4-Hydroxy-Phenyl)Ethanone, 1-(3-Allyl-4-Hydroxyphenyl)Ethanone, 1-[4-Hydroxy-3-(2-Propen-1-Yl)Phenyl]-Ethanone, 1-(4-Hydroxy-3-Prop-2-Enyl-Phenyl)Ethanone
- CAS Number:1132-05-4
- Formula:C₁₁H₁₂O₂
- MDL Number:MFCD00068015
2 Options Available Below
Product Details & Documents
Synonyms: 3′-Allyl-4′-hydroxyacetophenone, 1-(3-Allyl-4-Hydroxy-Phenyl)Ethanone, 1-(3-Allyl-4-Hydroxyphenyl)Ethanone, 1-[4-Hydroxy-3-(2-Propen-1-Yl)Phenyl]-Ethanone, 1-(4-Hydroxy-3-Prop-2-Enyl-Phenyl)Ethanone
- CAS Number:1132-05-4
- Formula:C₁₁H₁₂O₂
- MDL Number:MFCD00068015
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



