
2-Hydroxy-5-pyrimidineboronic acid pinacol ester 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1073354-84-3
:
Formula:C₁₀H₁₅BN₂O₃
MDL Number:MFCD09037497
:222.05 g/mol
Melting Pt:201…204 °C
:GHS07
Storage Temperature:Ambient