
4-Bromo-3,5-dimethyl-1H-pyrazole ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:3398-16-1
:
Formula:C₅H₇BrN₂
MDL Number:MFCD00005242
:175.03 g/mol
EINECS:222-263-2
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:121.0 to 125.0 deg-C