3-(1-Hydroxyethyl)benzophenone
Catalog # PROOMM0001.11
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
CAS Number:67173-18-6
Formula:C₁₅H₁₄O₂