Order Entry
Finland
ContactUsLinkComponent
Hydrochlorothiazide
Hydrochlorothiazide
Catalog # PROOBP186
CAS Number:  
Hydrochlorothiazide
Catalog # PROOBP186
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    200 mg
  • Seq:
    2530

Specifications

About this item

 :
EINECS:200-403-3
CAS Number:58-93-5
Formula:C₇H₈ClN₃O₄S₂
MDL Number:MFCD00051765