0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
Brass (alloy 260)
Brass (alloy 260), plate, Thickness: 6.35 mm (0.25 in)
Brass (alloy 260), plate, Thickness: 6.35 mm (0.25 in)
Brass (alloy 260), plate, Thickness: 6.35 mm (0.25 in)
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Brass (alloy 260), plate, Thickness: 6.35 mm (0.25 in)
Supplier:
Thermo Fisher Scientific
About this item
:
MDL Number:
MFCD00171255
Storage Temperature:
Ambient
Related Information
Documentation
SDS
msds_ALFA045182
Videos
Thermo Scientific Acroseal 25 Years
Documentation