Order Entry
Finland
ContactUsLinkComponent
Brass (alloy 260), foil, Thickness: 2.0 mm (0.08 in)
Brass (alloy 260), foil, Thickness: 2.0 mm (0.08 in)
Catalog # 45137.HB
CAS Number:  
Brass (alloy 260), foil, Thickness: 2.0 mm (0.08 in)
Catalog # 45137.HB
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    1886
  • Pk Info:
    300×300 mm

Specifications

About this item

 :
Storage Temperature:Ambient
MDL Number:MFCD00171255