Order Entry
Finland
Orders LinkContactUsLinkComponent
Buffer, HEPES solution (1 mol/L)

Buffer, HEPES solution (1 mol/L)

HEPES buffer solution (1 mol/l), Glass bottle, 500 mL
Catalog # EURLECM0180L
Supplier:   EUROCLONE
Buffer, HEPES solution (1 mol/L)
Catalog # EURLECM0180L
Supplier:  EUROCLONE
CAS Number:  
Buffer, HEPES solution (1 mol/L)

HEPES buffer solution (1 mol/l), Glass bottle, 500 mL

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    HEPES buffer solution (1 mol/l)

Specifications