
Diclofenac sodium salt ≥98.0% (by HPLC, total nitrogen)
Catalog # TCIAD2508-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Solubility in Methanol:almost transparency
Specification Test Results
About this item
:
:318.1 g/mol
Merck Index:14,03081
EINECS:239-346-4
UN:2811
ADR:6.1,III
:GHS06
CAS Number:15307-79-6
Formula:C₁₄H₁₀Cl₂NNaO₂
MDL Number:MFCD00082251