
Iminodiacetonitrile ≥98.0% (by total nitrogen basis)
Supplier: TCI
Certificates
About this item
CAS Number:628-87-5
:
Formula:C₄H₅N₃
MDL Number:MFCD00001887
:95.11 g/mol
EINECS:211-058-3
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:76.0 to 80.0 deg-C
- Solubility in hot Methanol:almost transparency