



Isopropyl chloroformate ∼30% ∼2 mol/L in toluene
: TCI
:108-23-6
:
:C₄H₇ClO₂
:MFCD00075068
:6°C
:Refrigerator
:203-563-2
:3286
:3,II
:GHS02
:GHS05
:GHS06
:GHS08
- Appearance:Colorless to Almost colorless clear liquid
- Concentration (Argentmetric titration):30.0 to 35.0 w/w%