Erlenmeyer flasks, wide neck, with screw cap, Azlon®
Supplier: SciLabware
Certificates
About this item
PP, translucent.
- Very good chemical resistance
- Resistant to heat up to 121 °C (for 20 minutes), prolonged use 100 °C
- Also sealable with standard ground joint stoppers (stoppers not supplied)
With blue, easy to read graduations. Autoclavable. The flasks feature a stopper bed and can also be sealed with standard ground joint polypropylene stoppers.