Magnetic stirring bars, cylindrical
Supplier: SEMADENI
Certificates
About this item
Magnetic stirrer bars with permanent magnetic core and white PTFE coating.
- Chemical resistance
- Smooth surface and round ends
- Temperature resistance upto 275 °C
Magnetic stirrer bars with permanent magnetic core and white PTFE coating.