Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tapes
Sealing tape
Sealing tape
Sealing tape
Supplier:
SEMADENI
Certificates
Expand image
Sealing tape
Supplier:
SEMADENI
About this item
Delivered on a plastic reel. Strength 0,075 mm.
Heat resistant up to 230 °C
Colour: White