Culture shake flasks
Supplier: BELLCO GLASS
Certificates
About this item
Flasks, made of type 1 class A borosilicate glass, with rigidly controlled baffle angles and depths to promote optimal oxygen transfer while reducing vortex formation.
- With break resistant lip to resist chipping
- Very good chemical resistance
- Very good temperature resistance
Designed for use with orbital shakers.