Order Entry
Finland
ContactUsLinkComponent
Serological pipettes, Shortie
Serological pipettes, Shortie
Catalog # BELC1224-05010
Supplier:  BELLCO GLASS
undefined
Serological pipettes, Shortie
Catalog # BELC1224-05010
Supplier:  BELLCO GLASS

Specifications

  • Colour code:
    Blue
  • Capacity:
    5 ml
  • Length:
    160 mm
  • Division:
    0,1 ml
  • Pk:
    18

Specifications

About this item

These serological pipettes are manufactured from type 1 borosilicate glass.

  • Shorter length facilitates use under biological hoods
  • Plain top promotes rapid, even flow (use rubber bulb to fill)
  • Calibrated to tip
  • Tempered tip; to deliver, blow-out
  • Will withstand repeated autoclaving