Cation exchange chromatography columns, HiTrap™ SP XL
: Cytiva
HiTrap SP XL are strong cation exchangers prepacked with SP Sepharose XL media, optimized for fast, convenient small-scale protein capture using ion exchange (IEX) chromatography.
- High binding capacities with high flow rates
- Convenient and affordable for fast, easy separations either alone or connected in series
- Designed for use with syringe, peristaltic pump, or chromatography system such as ÄKTA design
Q Sepharose XL and SP Sepharose XL resins have long chains of dextran coupled to a robust, 6% highly cross-linked agarose matrix. The dextran chains increase the exposure of the Q or SP charged groups, which results in very high loading capacities.
- Catalog No:
- 17-5160-01
- 17-5161-01
- Max. pressure:
- 5 bar
- 5 bar
- pH range:
- 4 - 13
- 4 - 13