Affinity chromatography columns, Ab SpinTrap™
Product Details & Documents
Ab SpinTrap columns are prepacked, single-use spin columns for rapid purification of monoclonal and polyclonal antibodies from serum and cell culture supernatants. Designed for small-scale purification of multiple samples in parallel.
- Rapid and convenient small-scale purification of IgG
- Protein G Sepharose High Performance in prepacked spin columns
- No filtration or centrifugation of different crude samples necessary for antibody purification
- Classic and crosslink protocols for protein enrichment (immunoprecipitation)
- Elution conditions formatted for both LC-MS and electrophoresis analysis
The spin columns are also suitable for use in antibody screening experiments. Ab SpinTrap columns are used together with a standard microcentrifuge and one purification run is performed in less than 20 min. Ab SpinTrap contains Protein G Sepharose High Performance, which has high protein binding capacity and is compatible with all commonly buffers used in antibody purification with immobilized Protein G media. For optimal performance in antibody purification, use Ab SpinTrap together with buffers prepared using Ab Buffer Kit.
Designed for small-scale purification of multiple samples in parallel. Ab SpinTrap is designed for efficient, small-scale purification of monoclonal and polyclonal antibodies from serum and cell culture supernatants. In addition, a protocol for immunoprecipitation is included.
Documents
28408347_DATA FILE.pdf
Datasheet
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications
- Particle Size:34 µm
- Capacity:100 µl
- Description:Ab SpinTrap™
- Seq:01
- Cat. No.:28-4083-47
- Binding capacity:> 1 mg IgG/column