About this item
Superdex prep grade is a preparative gel filtration medium with a composite matrix of dextran and agarose
- For gel filtration chromatography
- Gel filtration media providing exceptional resolution with short run times and good recovery
- BioProcess medium supported for industrial applications and well-established in approved processes
- Stable in all commonly used buffers
- Spherical particle form
Superdex prep grade is available in a range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development. As a BioProcess medium, Superdex prep grade meets industrial demands with security of supply and comprehensive technical and regulatory support.
Superdex 30 prep grade is designed for purification of molecules in the molecular weight range up to 10 000 MW, such as peptides, oligosaccharides and small proteins. Superdex 75 prep grade is recommended for the purification of recombinant proteins. (Fractionation range 3000 - 70000 Da). Superdex 200 prep grade is recommended for the purification of monoclonal antibodies. (Fractionation range 10000 - 600000 Da).
This matrix combines the excellent gel filtration properties of cross-linked dextran (Sephadex) with the physical and chemical stabilities of highly cross-linked agarose, to produce a separation medium with outstanding selectivity and high resolution. In addition, its low non-specific interaction permits high recovery of biological material. Together these properties make Superdex prep grade the first choice gel filtration media for all applications from laboratory to process scale.
Specifications
- pH range:3 - 12
- Matrix:Composite of cross-linked agarose and dextran
- Fractionation range:3 × 103 - 7 × 104 globular proteins; 5 × 102 - 3 × 104 dextrans
- Storage temperature:4 - 30 °C