Affinity chromatography columns, HiTrap™ MabSelect Xtra™
About this item
HiTrap™ MabSelect Xtra™ columns are prepacked with MabSelect Xtra™, a BioProcess medium for capture of monoclonal antbodies from large sample volumes.
- Increased dynamic binding capacity with smaller particle size and greater porosity than MabSelect™ medium
- MabSelect™ Xtra uses the same recombinant protein A ligand as MabSelect™
- Oriented coupling of recombinant protein A to the matrix via an engineered C-terminal cysteine enhances IgG binding capacity
- Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
The increased capacity of the medium is well suited for purification of Mabs from high-level expression feedstock.
3 Options Available Below
Product Details & Documents
HiTrap™ MabSelect Xtra™ columns are prepacked with MabSelect Xtra™, a BioProcess medium for capture of monoclonal antbodies from large sample volumes.
- Increased dynamic binding capacity with smaller particle size and greater porosity than MabSelect™ medium
- MabSelect™ Xtra uses the same recombinant protein A ligand as MabSelect™
- Oriented coupling of recombinant protein A to the matrix via an engineered C-terminal cysteine enhances IgG binding capacity
- Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
The increased capacity of the medium is well suited for purification of Mabs from high-level expression feedstock.
Documents
28408258_DATA FILE 1.pdf 28408258_DATA FILE.pdf 28408261_DATA FILE 1.pdf 28408261_DATA FILE.pdf
Datasheet
28408258_HAND BOOK 1.pdf 28408258_HAND BOOK.pdf 28408258_INSTRUCTIONS 1.pdf 28408258_INSTRUCTIONS 2.pdf 28408258_INSTRUCTIONS.pdf
Manual
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Additional Product Details
Specifications for Product Family
Specifications
- Catalog No:
- 28-4082-58
- 28-4082-60
- 28-4082-61
- Max. pressure:
- 5 bar
- 5 bar
- 5 bar
- pH range:
- 3 - 10
- 3 - 10
- 3 - 10
Specifications
Product Family Options
Product Information
- Column hardwareParticle SizeBed dimensionsCapacityFlow rateAvailabilityPrice
- 28-4082-58HiTrap™ MabSelect Xtra™Specifications:
More
Specifications
Cat. No.28-4082-58Max. pressure5 barpH range3 - 10 - 28-4082-60HiTrap™ MabSelect Xtra™HiTrap™ MabSelect Xtra™PP75 µm16×25 mm1×5 ml<20 ml/minLog in for availability1 * 1 Items1 050,00 €Specifications:
More
Specifications
Cat. No.28-4082-60Max. pressure5 barpH range3 - 10 - 28-4082-61HiTrap™ MabSelect Xtra™HiTrap™ MabSelect Xtra™PP75 µm16×25 mm5×5 ml<20 ml/minLog in for availability1 * 5 Items4 250,00 €Specifications:
More
Specifications
Cat. No.28-4082-61Max. pressure5 barpH range3 - 10