Coomassie® brilliant blue R-250, Ultrapure
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:6104-59-2
:
Formula:C₄₅H₄₄N₃NaO₇S₂
MDL Number:MFCD00041762
:825.98 g/mol
Melting Pt:174…180 °C
Storage Temperature:Ambient
EINECS:228-060-5
Specification Test Results
- Dye content (dry basis):≥65 %
- Em (550 nm, water, dry basis):≥33000
- Loss on drying:≤5.0%
- Solubility (0.020 g/l, water):Pass