Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ponceau S
Ponceau S for electrophoresis
Ponceau S for electrophoresis
Ponceau S for electrophoresis
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Ponceau S for electrophoresis
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
6226-79-5
:
Formula:
C₂₂H₁₂N₄Na₄O₁₃S₄
MDL Number:
MFCD00003892
:
760.56 g/mol
EINECS:
228-319-2
:
GHS07
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAJ60744
Videos
Thermo Scientific Acroseal 25 Years
Documentation