Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Kanamycin sulphate
Kanamycin sulphate
Kanamycin sulphate
Kanamycin sulphate
Supplier:
Thermo Fisher Scientific
Certificates
Expand image
Kanamycin sulphate
Supplier:
Thermo Fisher Scientific
Certificates
About this item
CAS Number:
25389-94-0
:
Formula:
C₁₈H₃₈N₄O₁₅S
MDL Number:
MFCD00070253
:
582.6 g/mol
Merck Index:
14,5281
EINECS:
246-933-9
:
GHS08
Storage Temperature:
Ambient
Related Information
Documentation
Certificates
Search by Lot Number
SDS
msds_ALFAJ61272
Application Notes
Organic Building Blocks and Intermediates
Videos
Thermo Scientific Acroseal 25 Years
Documentation