Ampicillin sodium salt
About this item
CAS No.: 69-52-3 Molecular Formula: C16H18N3NaO4S Formula Weight: 371.39 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 215° Solubility: Freely soluble in water. Sparingly soluble in acetone Beilstein Reference: 4119211 Merck Reference: 14,586 MDL No.: MFCD00064313
- CAS Number:69-52-3
- Formula:C₁₆H₁₈N₃NaO₄S
- MDL Number:MFCD00064313
- Molecular Weight:371.39 g/mol
- Merck Index:14,586
- Melting Pt:215 °C
- Storage Temperature:Refrigerator
- EINECS:200-708-1
2 Options Available Below
Product Details & Documents
CAS No.: 69-52-3 Molecular Formula: C16H18N3NaO4S Formula Weight: 371.39 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 215° Solubility: Freely soluble in water. Sparingly soluble in acetone Beilstein Reference: 4119211 Merck Reference: 14,586 MDL No.: MFCD00064313
- CAS Number:69-52-3
- Formula:C₁₆H₁₈N₃NaO₄S
- MDL Number:MFCD00064313
- Molecular Weight:371.39 g/mol
- Merck Index:14,586
- Melting Pt:215 °C
- Storage Temperature:Refrigerator
- EINECS:200-708-1
Documents
Product Family Options
Product Information
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



