Order Entry
Finland
ContactUsLinkComponent
Hydrogen chloride ∼4 mol/L in 1,4-dioxane
Flammable
Corrosive
Irritant
Health
Hydrogen chloride ∼4 mol/L in 1,4-dioxane
Supplier:  TCI
Certificates
undefined
Flammable
Corrosive
Irritant
Health
Hydrogen chloride ∼4 mol/L in 1,4-dioxane
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Concentration (Neutralization titration):
    14.0 to 15.4 w/w%

Specification Test Results