Order Entry
Finland
ContactUsLinkComponent
1,2-Difluoro-4-iodobenzene ≥98.0% (by GC) stabilised
Irritant
1,2-Difluoro-4-iodobenzene ≥98.0% (by GC) stabilised
Catalog # TCIAD2560-10G
Supplier:  TCI
CAS Number:  
undefined
Irritant
1,2-Difluoro-4-iodobenzene ≥98.0% (by GC) stabilised
Catalog # TCIAD2560-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green clear liquid
  • Purity (GC):
    min. 98.0 %

Specification Test Results

About this item

 :
 :239.99 g/mol
Boiling Pt:177 °C (10 -rr)
Density:1,99 g/cm³ (20 °C)
Flash Pt:73°C
 :Stabilised with copper chip
 :GHS07
Storage Temperature:Refrigerator
CAS Number:64248-58-4
Formula:C₆H₃F₂I
MDL Number:MFCD00044630