Order Entry
Finland
ContactUsLinkComponent
Coomassie® brilliant blue R-250, Laboratory Reagent, J.T.Baker®
Coomassie® brilliant blue R-250, Laboratory Reagent, J.T.Baker®
Catalog # F792-01
Supplier:  Avantor
CAS Number:  
undefined
Coomassie® brilliant blue R-250, Laboratory Reagent, J.T.Baker®
Catalog # F792-01
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :825.98 g/mol
Melting Pt:174…180 °C
Storage Temperature:Ambient
EINECS:228-060-5
CAS Number:6104-59-2
Formula:C₄₅H₄₄N₃NaO₇S₂
MDL Number:MFCD00041762