Bromocresol green sodium salt ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:62625-32-5
:1007478-52-5
Formula:C₂₁H₁₃Br₄NaO₅S
MDL Number:MFCD00148898
Merck Index:14,1386
Melting Pt:230 °C (dec.)
:720 g/mol
Storage Temperature:Ambient
EINECS:263-657-4