
1-Butyl-3-methylimidazolium dicyanamide ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:448245-52-1
:MFCD17976576
Formula:C₁₀H₁₅N₅
MDL Number:MFCD07784457
:205.26 g/mol
Density:1.06 g/cm³ (20 °C)
:GHS07
Storage Temperature:Ambient