Guanidinium thiocyanate for molecular biology
:593-84-0
:
:CH₅N₃·CHNS
:MFCD00013027
:118.16 g/mol
:118…122 °C
:1.29
:209-812-1
:GHS07
:Ambient
- Appearance (Colour):White to off-white
- Form:Crystalline powder
- Assay (Titration):99.0% min.
- DNase:None detected
- Protease activity:None detected
- RNase:None detected
- UV/Vis A280 (70%, water):0.8
- UV/Vis A300 (6M, water):0.1
- UV/Vis A410 (6M, water):0.1