Order Entry
Finland
ContactUsLinkComponent
4-Bromobenzylamine hydrochloride ≥97.0% (by total nitrogen and titration analysis)
Irritant
4-Bromobenzylamine hydrochloride ≥97.0% (by total nitrogen and titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
4-Bromobenzylamine hydrochloride ≥97.0% (by total nitrogen and titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Purity (Argentmetric Titration):
    min. 97.0 %

Specification Test Results

Frequently Bought Together