Order Entry
Finland
ContactUsLinkComponent
Nickel copper alloy (67:33 w%), alloy 400, foil, Thickness: 0.51 mm (0.02 in)
Nickel copper alloy (67:33 w%), alloy 400, foil, Thickness: 0.51 mm (0.02 in)
Catalog # 45148.HB
CAS Number:  
Nickel copper alloy (67:33 w%), alloy 400, foil, Thickness: 0.51 mm (0.02 in)
Catalog # 45148.HB
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    1886
  • Pk Info:
    300×300 mm

Specifications

About this item

 :
 :GHS08
 :GHS07
Storage Temperature:Ambient