Order Entry
Finland
ContactUsLinkComponent
Nickel sulphide ≥99% (metals basis)
Health
Irritant
Nickel sulphide ≥99% (metals basis)
Catalog # 45498.06
CAS Number:  
Health
Irritant
Nickel sulphide ≥99% (metals basis)
Catalog # 45498.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

CAS Number:12035-51-7
 :
Formula:NiS
MDL Number:MFCD00016266
 :240.21 g/mol
UN:3077
ADR:9,III
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:12035-72-2
 :16812-54-7
Formula:Ni₃S₂
MDL Number:MFCD00217044