
Allyl methyl sulphide ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:10152-76-8
:
Formula:C₄H₈S
MDL Number:MFCD00008657
:88.17 g/mol
Boiling Pt:92°C
Flash Pt:18°C
EINECS:233-422-0
UN:1993
ADR:3,II
:GHS02
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %